Crystal violet solution (Hucker Formulation) for Gram staining

Supplier: Ricca Chemical
Danger

R3240000-500A R3240000-1A R3240000-120A R3240000-4A
RC324016EA 106 USD
RC324016 RC324032 RC32404 RC3240-1
Crystal violet solution (Hucker Formulation) for Gram staining
Crystal Violet

Gentian Violet, Hucker Formulation, for Gram Staining

Formula: C₂₅H₃₀ClN₃
MDL Number: MFCD00011750
CAS Number: 548-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR