Fluoride ISE Standards, Ricca Chemical
Supplier: Ricca Chemical
3161.5-32
RC3161.5-32EA
106
USD
RC3161.5-32
Fluoride ISE Standards, Ricca Chemical
Standards
Anionic Standards
Fluoride Standards
Fluoride standard, 1 mL = 0.0005 mg F-, 0.5 ppm F-, premixed with TISAB.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)