Iron(III) chloride 40% (w/v) in aqueous solution analytical reagent
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
Iron trichloride, Ferric chloride
R3127000-500A
R3127000-20F
RC3127-16EA
164
USD
RC3127-16
RCR3127000-20F
Iron(III) chloride 40% (w/v) in aqueous solution analytical reagent
Iron (III) chloride
Formula:
FeCl₃ |
MDL Number:
MFCD00011005 CAS Number: 7705-08-0 EINECS: 231-729-4 UN: 2582 ADR: 8,III |
Specification Test Results
Appearance | Clear, brown, odorless |
Assay at 20 °C (traceable to NIST SRM 136) | 40.0 ± 0.1 % (w/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...