Iron(III) chloride solution R1

Supplier: Ricca Chemical
Danger

Synonyms: Iron trichloride, Ferric chloride

R3105000-500B
RC3105-16EA 112 USD
RC3105-16
Iron(III) chloride solution R1
Iron (III) chloride

Ferric chloride solution R1

Formula: FeCl₃
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.1000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011005
CAS Number: 7705-08-0
UN: 2582
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear to slightly hazy, bright orange, odorless
Assay at 20 °C (traceable to NIST SRM 136) 105 ± 2 g/L FeCl3.6H2O


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR