EDTA disodium salt solution 0.1 mol/l in aqueous solution titrant

Supplier: Ricca Chemical

R2730000-4A R2730000-1A R2730000-20F
RC27301EA 270 USD
RC27301 RC273032 RC27305
EDTA disodium salt solution 0.1 mol/l in aqueous solution titrant
EDTA disodium salt

Standardized at 25°C against chemicals whose certification is traceable to NIST.

Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
Melting Pt: 255 °C
Storage Temperature: Ambient
MDL Number: MFCD00150037
CAS Number: 6381-92-6
EINECS: 205-358-3
Merck Index: 13,03543

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 915) 0.1000 ± 0.0001 M at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR