EDTA disodium salt 0.0500 M (M/20) in aqueous solution ASTM D 2036 D titrant

Supplier: Ricca Chemical

Synonyms: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt

2705-16
RC2705-16EA 55 USD
RC2705-16
EDTA disodium salt 0.0500 M (M/20) in aqueous solution ASTM D 2036 D titrant
EDTA disodium salt
Formula: C₁₀H₁₄N₂Na₂O₈
MW: 336.21 g/mol
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0090 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00070672
CAS Number: 139-33-3

Order Now

Specification Test Results

Assay 0.05 ± 0.00001 M at 20 °C
NIST SRM 915

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR