EDTA disodium salt 0.0100 M (M/100) in aqueous solution

Supplier: Ricca Chemical

R2700000-4A R2700000-10F R2700000-1A R2700000-20F R2700000-500A
RC27001EA 114 USD
RC27001 RC270025 RC270032 RC27005 RC2700-16
EDTA disodium salt 0.0100 M (M/100) in aqueous solution
EDTA disodium salt
Formula: C₁₀H₁₄N₂Na₂O₈
Melting Pt: 255 °C
Storage Temperature: Ambient
MDL Number: MFCD00070672
CAS Number: 139-33-3
EINECS: 205-358-3

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 915) 0.01000 ± 0.00001 M at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR