EDTA disodium salt 0.0100 M (M/100) in aqueous solution
Supplier: Ricca Chemical
Synonyms:
Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt
R2700000-4A
R2700000-10F
R2700000-1A
R2700000-20F
R2700000-500A
RC27001EA
114
USD
RC27001
RC270025
RC270032
RC27005
RC2700-16
EDTA disodium salt 0.0100 M (M/100) in aqueous solution
EDTA disodium salt
Formula:
C₁₀H₁₄N₂Na₂O₈ MW: 336.21 g/mol Melting Pt: 255 °C Storage Temperature: Ambient |
MDL Number:
MFCD00070672 CAS Number: 139-33-3 EINECS: 205-358-3 |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 915) | 0.01000 ± 0.00001 M at 20.0 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...