EDTA disodium salt solution 0.00100 M (M/1000) titrant
Supplier: Ricca Chemical
R2692000-500A
R2692000-1A
RC2692-16EA
43
USD
RC2692-16
RC2692-32
EDTA disodium salt solution 0.00100 M (M/1000) titrant
EDTA disodium salt
1 ml = 0.1 mg CaCO3 0.04 mg Ca
Formula:
C₁₀H₁₄N₂Na₂O₈·2H₂O Boiling Pt: 100 °C Melting Pt: 0.0 °C Density: 1.0010 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00150037 CAS Number: 6381-92-6 |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 915) | 0.001000 ± 0.000005 M at 20.0 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...