EDTA disodium salt 5% in sodium hydroxide solution for ammonia nitrogen determination
Supplier: Ricca Chemical
R2669000-500A
R2669000-1A
RC2669-16EA
90
USD
RC2669-16
RC2669-32
EDTA disodium salt 5% in sodium hydroxide solution for ammonia nitrogen determination
EDTA disodium salt
Formula:
C₁₀H₁₄N₂Na₂O₈·2H₂O Boiling Pt: ~ 100 °C Melting Pt: ~ 0 °C Density: 1.0200 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00150037 CAS Number: 6381-92-6 |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 915) | 5.00 ± 0.04 % (w/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...