EDTA disodium salt 0.1% (w/v) in 5% (v/v) alcohol USP Test Solution (TS)

Supplier: Ricca Chemical
Danger

Synonyms: Ethylenedinitrilotetra-acetic acid disodium salt dihydrate, Kestranal® 2S 2 H2O, Edetic acid disodium salt dihydrate, Titriplex III, Disodium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenediaminetetra-acetic acid disodium salt dihydrate, Kestranal® 2S dihydrate, Edetate disodium dihydrate

R2664000-500A
RC2664-16EA 142 USD
RC2664-16
EDTA disodium salt 0.1% (w/v) in 5% (v/v) alcohol USP Test Solution (TS)
EDTA disodium salt
Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
MW: 372.24 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 0.9905 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00150037
CAS Number: 6381-92-6

Order Now

Specification Test Results

Appearance Clear, colorless, odorless to slight alcohol odor
Assay at 20 °C (traceable to NIST SRM 915) 1.00 ± 0.01 g/L

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR