Dextrose solution 0.5% (w/v) in hydrochloric acid 0.25% for D.E. test on syrup

Supplier: Ricca Chemical
Danger

Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose

R2548000-4A
RC25481EA 517 USD
RC25481
Dextrose solution 0.5% (w/v) in hydrochloric acid 0.25% for D.E. test on syrup
D(+)-Glucose
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0010 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00148912
CAS Number: 50-99-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless
Certified Concentration 0.500 ± 0.001 % (w/v) Dextrose

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR