Reagent alcohol 50% (v/v) in water (denatured with methanol and isopropanol)

Supplier: Ricca Chemical
Danger

Synonyms: Reagent alcohol

2546.50-16
RC2546.50-16EA 50 USD
RC2546.50-16
Reagent alcohol 50% (v/v) in water (denatured with methanol and isopropanol)
Alcohol
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR