Ethanol 50% (v/v) analytical reagent

Supplier: Ricca Chemical
Danger

Synonyms: Ethyl alcohol

R2546500-4A
RC2546.50-1CS 2371 USD
RC2546.50-1
Ethanol 50% (v/v) analytical reagent
Ethanol

Clear, colorless liquid. Denatured with 2 to 3% methanol and 2 to 3% 2-propanol.


  • Total alcohol: ~ 50%
  • Ethyl alcohol: 23-50%
  • Denaturant: 1-3% methyl alcohol and 1-3% isopropyl alcohol

Formula: C₂H₅OH
MW: 46.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,III
Merck Index: 13,03795

Order Now

Specification Test Results

Appearance Clear, colorless, alcohol odor
Assay 50.0 ± 1.0 % (v/v)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR