Crystal violet 0,2% (w/v) in glacial acetic acid indicator for nonaqueous titrations

Supplier: Ricca Chemical

Synonyms: Methylrosanilide chloride, Basic violet 3, Viocid, N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanamonium chloride, Violet crystal, Methyl violet 10B, Dimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-ylidene)ammonium chloride, CI 42555, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride, Hexamethyl violet, Hexamethyl-p-rosaniline chloride, Vianin, Gentian violet

R2480000-120C
RC2480-4EA 457 USD
RC2480-4
Crystal violet 0,2% (w/v) in glacial acetic acid indicator for nonaqueous titrations
Crystal Violet
Formula: C₂₅H₃₀ClN₃
MW: 407.99 g/mol
MDL Number: MFCD00011750
CAS Number: 548-62-9
UN: 3077
ADR: 9 III
Merck Index: 13,04406

Order Now

Specification Test Results

Appearance Dark purple, acetic acid odor
Suitability for Use Yellow (pH 2.0)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR