Copper(II) sulfate 0.0100 M (M/100)

Supplier: Ricca Chemical

Synonyms: Cupric sulphate, Cupric sulfate

2347-32
RC2347-32EA 47 USD
RC2347-32
Copper(II) sulfate 0.0100 M (M/100)
Copper (II) sulphate

THIS PRODUCT IS CERTIFIED AS A REAGENT FOR THE FOLLOWING TEST METHODS:
Ref - TAPPI Method - Glue in paper (qualitative and quantitative determination)

Formula: CuSO₄
MW: 159.61 g/mol
MDL Number: MFCD00010981
CAS Number: 7758-98-7
UN: 3288
ADR: 6.1,III

Order Now

Specification Test Results

Assay 0.01 ± 0.00001 M at 20 °C
NIST SRM 136

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR