Citric acid 10% (w/v) in aqueous solution

Supplier: Ricca Chemical
Danger

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid

2120-32
RC2120-32EA 74 USD
RC2120-32
Citric acid 10% (w/v) in aqueous solution
Citric acid

Manufactured from USP grade material.

Formula: HOC(COOH)(CH₂COOH)₂
MW: 192.13 g/mol
Boiling Pt: 310
Melting Pt: ~ 0 °C
Density: 1.0405 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011669
CAS Number: 77-92-9
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d60f5a9b8e6c6dfb7eda04e1a1d5d6d971f92444&vid=caf4f9effc7fd22b942f7f92be770b06e40637a4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 10 ± 0.03 % (w/v)
NIST SRM 84


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR