Cerium(IV) sulfate 0.250 N (N/4) in sulfuric acid 2N titrant

Supplier: Ricca Chemical

Synonyms: Ceric sulfate, Cerium disulfate, Cerium disulphate, Ceric sulphate, Cerium(IV) sulfate, Cerium(IV) sulphate

1907-1
RC1907-1EA 680 USD
RC1907-1
Cerium(IV) sulfate 0.250 N (N/4) in sulfuric acid 2N titrant
Cerium (IV) sulphate
Formula: Ce(SO₄)₂
MW: 332.24 g/mol
MDL Number: MFCD00148852
CAS Number: 13590-82-4
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Assay 0.25 ± 0.0001 N at 20 °C
NIST SRM 40

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR