Calcium sulfate solution R, saturated analytical reagent

Supplier: Ricca Chemical

Synonyms: Calcium sulphate (anhydrous) anhydrite, Gypsum, Calcium sulfate (anhydrous) anhydrite

R1823000-120A R1823000-500A
RC1823-4EA 385 USD
RC1823-4 RC1823-16
Calcium sulfate solution R, saturated analytical reagent
Calcium sulphate
Formula: CaO₄S
MW: 136.14 g/mol
Melting Pt: 1350
Storage Temperature: Ambient
MDL Number: MFCD00010912
CAS Number: 7778-18-9
EINECS: 231-900-3
Merck Index: 13,01711

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Calcium Present
Sulfate Present


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR