Calcium hydroxide saturated solution in water USP Test Solution (TS)

Supplier: Ricca Chemical

Synonyms: Slaked lime

R1785000-4A R1785000-500A R1785000-20F R1785000-1A
RC1785-1EA 361 USD
RC1785-1 RC1785-16 RC1785-5 RCR1785000-1A
Calcium hydroxide saturated solution in water USP Test Solution (TS)
Calcium hydroxide

Multicompendial test reagent.


  • USP Test SolutionCalcium Hydroxide TS
  • TAPPI Reagent 5.9 Lime Water Saturated Solution
  • EP Reagent Calcium hydroxide solution R

Formula: Ca(OH)₂
Boiling Pt: ~100 °C
Melting Pt: ~0 °C
Storage Temperature: Ambient
MDL Number: MFCD00010901
CAS Number: 1305-62-0
UN: 3262
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR