Calcium chloride 7.5% (w/v) in aqueous solution USP Test Solution (TS)
Supplier: Ricca Chemical
R1783000-500A
R1783000-1A
RC1783-16EA
93
USD
RC1783-16
RC1783-32
Calcium chloride 7.5% (w/v) in aqueous solution USP Test Solution (TS)
Calcium chloride
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 915) | 7.50 ± 0.01 % (w/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...