Calcium chloride 2,75% (w/v) in aqueous solution

Supplier: Ricca Chemical
Danger

Synonyms: Chloro calcium, Calcium dichloride, CaCl2

R1720000-500A R1720000-1A R1720000-4A
RC172016EA 55 USD
RC172016 RC172032 RC1720-1
Calcium chloride 2,75% (w/v) in aqueous solution
Calcium chloride
Formula: CaCl₂
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0170 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00010903
CAS Number: 10043-52-4

Order Now

Specification Test Results

% (w/v) (traceable to NIST SRM 915) 2.750 ± 0.005 % (w/v)
Appearance Clear, colorless, odorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR