Calcium chloride 2,75% (w/v) in aqueous solution
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
Chloro calcium, Calcium dichloride, CaCl2
R1720000-500A
R1720000-1A
R1720000-4A
RC172016EA
55
USD
RC172016
RC172032
RC1720-1
Calcium chloride 2,75% (w/v) in aqueous solution
Calcium chloride
Formula:
CaCl₂ Boiling Pt: ~ 100 °C Melting Pt: ~ 0 °C Density: 1.0170 g/cm³ Storage Temperature: Refrigerator |
MDL Number:
MFCD00010903 CAS Number: 10043-52-4 |
Specification Test Results
% (w/v) (traceable to NIST SRM 915) | 2.750 ± 0.005 % (w/v) |
Appearance | Clear, colorless, odorless |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...