R1489000-4A
R1489000-1A
R1490000-4A
R1490000-1A
R1490000-500A
R1492000-4A
R1492000-1A
R1493000-4A
R1493000-20F
R1495000-4A
R1495000-1A
R1495000-20F
R1498000-4A
R1498000-1A
R1498000-500A
R1500000-4A
R1500000-10F
R1500000-1A
R1500000-20F
R1505000-4A
R1505000-1A
R1510000-4A
R1510000-1A
R1540000-4A
R1540000-1A
R1540000-500A
R1550000-4A
R1550000-500A
R1550000-10F
R1550000-1A
R1550000-20F
R1489000-500A
R1492000-500A
R1493000-500A
R1493000-1A
R1495000-500A
R1500000-500A
R1505000-500A
R1510000-500A
R1550000-4F
R1492000-20F
R1500000-4F
1563-16
1485-1
1485-16
1485-32
R1492000-20PK
R1493000-20PK
R1489000-20F
R1492000-10F
R1493000-10F
R1495000-10F
R1495000-20PK
R1498000-20F
R1505000-10F
R1505000-20F
R1510000-10F
R1540000-10F
R1540000-20F
R1565000-1A
R1565000-20F
R1565000-4A
R1565000-500A
RC1489-1EA
244
USD
RC1489-1
RC1489-32
RC1490-1
RC1490-32
RC149016
RC1492-1
RC1492-32
RC1493-1
RC1493-5
RC1495-1
RC1495-32
RC1495-5
RC1498-1
RC1498-32
RC149816
RC15001
RC150025
RC150032
RC15005
RC1505-1
RC1505-32
RC1510-1
RC1510-32
RC1540-1
RC1540-32
RC154016
RC15501
RC155016
RC155025
RC155032
RC15505
RC1489-16
RC149216
RC1493-16
RC1493-32
RC1495-16
RC150016
RC1505-16
RC1510-16
RC1550-1CT
RC1492-5
RC1500-1CT
RC1563-16
RC1485-1
RC1485-16
RC1485-32
RCR149200020PK
RCR149300020PK
RCR1489000-20F
RCR1492000-10F
RCR1493000-10F
RCR1495000-10F
RCR149500020PK
RCR1498000-20F
RCR1505000-10F
RCR1505000-20F
RCR1510000-10F
RCR1540000-10F
RCR1540000-20F
RCR1565000-1A
RCR1565000-20F
RCR1565000-4A
RCR1565000500A
Buffers, Reference standards, Acidic to Neutral, Ricca Chemical
Buffers
pH Calibration Buffers
Colorless pH buffer solutions are certified to ±0.01 pH at 25°C
- Range of packaging is available
Each 500 ml plastic bottle is labeled with the certified pH value, the NIST SRM number and a pH/temperature conversion.
Certifications: Standardized against solutions whose certification is traceable to the National Institute of Standards and Technology. Meet USP requirements.