Acetic acid 1.00 N

Supplier: Ricca Chemical
Danger

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

150-16
RC150-16EA 34 USD
RC150-16
Acetic acid 1.00 N
Acetic acid
Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0084 g/cm³
Flash Pt: 39 °C (AcOH)
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 1 ± 0.002 N at 20 °C
NIST SRM 84

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR