Buffer Assortment Pack, RICCA Chemical Company
Supplier: Ricca Chemical
1480-16
RC1480-16EA
72
USD
RC1480-16
Buffer Assortment Pack, RICCA Chemical Company
Buffers
pH Calibration Buffers
Buffer assortment pack includes buffers with pH 4, 7 and 10
All buffers are also available separately.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...