Acetic acid 50% (v/v) in aqueous solution (1 + 1)

Supplier: Ricca Chemical

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

140-5
RC140-5EA 442 USD
RC140-5
Acetic acid 50% (v/v) in aqueous solution (1 + 1)
Acetic acid
Formula: H₃CCOOH
MW: 60.05 g/mol
MDL Number: MFCD00036152
CAS Number: 64-19-7
UN: 2790
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 50 ± 0.05 % (v/v)
NIST SRM 84

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR