Bromocresol green 0.04% (w/v) in aqueous solution indicator

Supplier: Ricca Chemical

R1200000-500A R1200000-1A R1200000-120A
RC120016EA 61 USD
RC120016 RC120032 RC12004
Bromocresol green 0.04% (w/v) in aqueous solution indicator
Bromocresol green
Formula: C₂₁H₁₄Br₄O₅S
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0010 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00005874
CAS Number: 76-60-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12b4d4b3d992f8e86746537bf756af442752c853&vid=553ef1836a5b191a43558f8550e78cf2e424446e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Blue-green, odorless
Suitability for Use Blue (pH 5.4) - Green (pH 4.5) - Yellow (pH 4.0)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR