6-Carboxyfluorescein

Supplier: BACHEM AMERICAS INC

Synonyms: 3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-6-carboxylic acid, 6-FAM

4045142.0100 4045142.0500
Q-2665.0100BAEA 134.03 USD
Q-2665.0100BA Q-2665.0500BA
6-Carboxyfluorescein
6-Carboxyfluorescein

Sequence: 6-Carboxy-fluorescein
Synonym(s): 6-FAM#2-(6-Hydroxy-3-oxo-3H-xanthen-9-yl)-terephthalic acid


Bachem also offers the coumarin derivative Fmoc-3-(7-methoxy-coumarin-4-yl)-Ala-OH (B-3740), the (7-Hydroxycoumarin-4-yl)ethylglycine derivatives B-4205 and F-4270, and a range of Nε-labeled lysine derivatives (please see the subfamily "Nε-Labeled Lysine").


Widely used green-fluorescent labeling dye.

Formula: C₂₁H₁₂O₇
MW: 376.32 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00036873
CAS Number: 3301-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR