6-Carboxyfluorescein
Supplier: BACHEM AMERICAS INC
Synonyms:
3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-6-carboxylic acid, 6-FAM
4045142.0100
4045142.0500
Q-2665.0100BAEA
134.03
USD
Q-2665.0100BA
Q-2665.0500BA
6-Carboxyfluorescein
6-Carboxyfluorescein
Sequence: 6-Carboxy-fluorescein
Synonym(s): 6-FAM#2-(6-Hydroxy-3-oxo-3H-xanthen-9-yl)-terephthalic acid
Bachem also offers the coumarin derivative Fmoc-3-(7-methoxy-coumarin-4-yl)-Ala-OH (B-3740), the (7-Hydroxycoumarin-4-yl)ethylglycine derivatives B-4205 and F-4270, and a range of Nε-labeled lysine derivatives (please see the subfamily "Nε-Labeled Lysine").
Widely used green-fluorescent labeling dye.
Formula:
C₂₁H₁₂O₇ MW: 376.32 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00036873 CAS Number: 3301-79-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...