5-Carboxyfluorescein

Supplier: BACHEM AMERICAS INC

Synonyms: Spiro(isobenzofuran-1(3H),9'-(9H)xanthene)-5-carboxylic acid, 3',6'-dihydroxy-3-oxo-, 5-FAM, 3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-5-carboxylic acid, 5-Carboxyfluorescein hydrate

4045141.0100 4045141.0500
Q-2660.0100BAEA 134.03 USD
Q-2660.0100BA Q-2660.0500BA
5-Carboxyfluorescein
5-Carboxyfluorescein

Sequence: 5-Carboxy-fluorescein
Synonym(s): 5-FAM#4-(6-Hydroxy-3-oxo-3H-xanthen-9-yl)-isophthalic acid


Bachem also offers the coumarin derivative Fmoc-3-(7-methoxy-coumarin-4-yl)-Ala-OH (B-3740), the (7-Hydroxycoumarin-4-yl)ethylglycine derivatives B-4205 and F-4270, and a range of Nε-labeled lysine derivatives (please see the subfamily "Nε-Labeled Lysine").


Widely used green-fluorescent labeling dye.

Formula: C₂₁H₁₂O₇.xH₂O
Storage Temperature: Freezer
MDL Number: MFCD00036874
CAS Number: 76823-03-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR