EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride)

Supplier: BACHEM AMERICAS INC

4031236.0025 4031236.0100
Q-1955.0025BAEA 275.21 USD
Q-1955.0025BA Q-1955.0100BA
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride)
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride

Synonym(s): EDAC · HCl#EDC · HCl#N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide · HCl#Water-soluble Carbodiimide#WSC


A water-soluble coupling reagent for solution synthesis. WSCD has also been employed in SPPS in aqueous systems.

Formula: C₈H₁₇N₃·HCl
Storage Temperature: Refrigerator
MDL Number: MFCD00012503
CAS Number: 25952-53-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25d8bf2864bcf21ffe6dcca6f22388d1fe427e22&vid=9080dd6b66a1622ef32a2da2138f85e44cc881b8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR