D-Biotin-N-succinimidyl ester
Supplier: BACHEM AMERICAS INC
Synonyms:
Biotin-NHS, 2,5-Dioxopyrrolidin-1-yl 5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoate, (+)-Biotin N-hydroxysuccinimide ester, D-Biotinyl-OSu, NHS-Biotin, Biotin N-hydroxysuccinimide ester, N-(D-Biotinyloxy)succinimide
Sequence: D-Biotinyl-OSu
Synonym(s): BNHS#Biotin-NHS
For DOTA(OtBu)₃ see Q-2525.
The activated biotin derivative Biotin-OSu is used for labeling the amino groups of amino acids, peptides, and proteins. When conducting the biotinylation at pH 6.5, N-terminal amino groups can be selectively tagged in the presence of lysine.
Formula:
C₁₄H₁₉N₃O₅S Storage Temperature: Refrigerator |
MDL Number:
MFCD00078531 CAS Number: 35013-72-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...