2-Acetamido-2-deoxy-β-D-glucopyranosyl L-asparagine
Supplier: BACHEM AMERICAS INC
Synonyms:
β-D-GlcNAc-(1→N)-Asn, Ng-(2-Acetamido-2-deoxy-b-D-gluco-pyranosyl)-L-asparagine, 2-Acetamido-1-β-(L-aspartamido)-1 ,2-dideoxy-D-glucose, H-Asn(GlcNAc-β-D)-OH
4027708.0250
4027708.1000
Q-1725.0250BAEA
1340.71
USD
Q-1725.0250BA
Q-1725.1000BA
2-Acetamido-2-deoxy-β-D-glucopyranosyl L-asparagine
2-Acetamido-2-deoxy-β-D-glucopyranosyl L-asparagine
Sequence: H-Asn(GlcNAc-β-D)-OH
For isoasparagine derivatives please see the subfamily "Isoasparagine" filed under "Aspartic Acid".
Synonym(s): AADG#Aspartylglucosamine#N-γ-(2-Acetamido-2-deoxy-β-D-gluco-pyranosyl)-L-asparagine
Formula:
C₁₂H₂₁N₃O₈ Storage Temperature: Refrigerator |
MDL Number:
MFCD02259695 CAS Number: 2776-93-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...