Sulfo-SMCC (3-Sulfo-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt), Premium Grade, Pierce™

Supplier: 0000013509
Warning

PG82085 PG82086
PIPG82085EA 836.92 USD
PIPG82085 PIPG82086
Sulfo-SMCC (3-Sulfo-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt), Premium Grade, Pierce™
Sulfo-SMCC (3-Sulfo-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt)

Thermo Scientific Pierce Premium Grade Sulfo-SMCC is our highest quality formulation of amine-to-sulfhydryl crosslinker, specially characterized for applications where product integrity and risk minimization are paramount.


  • Sulfo-SMCC—a water-soluble amine-to-sulfhydryl crosslinker with a stabilizing cyclohexane spacer arm
  • High quality—identity and purity confirmed by several tests, including quantitative NMR
  • Product integrity—enhanced level of testing and characterization compared to standard grade
  • Lot retention—ample supply of past lots retained to ensure future process testing
  • Change management—Change Control Notification (CCN) service
  • Consistent manufacture—batch-specific manufacturing documentation review


Compared to the standard grade product, Premium Grade Sulfo-SMCC provides more clearly defined quality and product support by including (a) increased analytical testing and product characterization, (b) greater batch-specific information and quality assurance review, (c) extensive lot sample retention and (d) change control notification. Sulfo-SMCC is a water-soluble, non-cleavable and membrane-impermeable amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a medium-length cyclohexane spacer arm (8.3 angstroms). NHS esters react with primary amines at pH 7-9 to form stable amide bonds. Maleimides react with sulfhydryl groups at pH 6.5-7.5 to form stable thioether bonds. The maleimide groups of Sulfo-SMCC are unusually stable up to pH 7.5 because of the cyclohexane bridge in the spacer arm. Because it contains the hydrophilic sulfonyl moiety, Sulfo-SMCC is soluble up to nearly 10 mM in water and many commonly used buffers, thus avoiding the use of organic solvents which may perturb protein structure.


Caution: For Research Use Only. Not for use in diagnostic procedures.

Formula: C₁₆H₁₈N₂O₉S
Storage Temperature: Freezer
MDL Number: MFCD00054978
CAS Number: 92921-24-9

Order Now

Specification Test Results

Product Line Pierce™
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Maleimide, Sulfo-NHS Ester
Spacer Short
Form Powder
Quantity 100 mg/1 g
Type Crosslinker
Solubility Water


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR