EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride), Premium Grade, Pierce™

Supplier: 0000013509
Warning

PG82073 PG82079
PIPG82073EA 650.4 USD
PIPG82073 PIPG82074 PIPG82079
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride), Premium Grade, Pierce™
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride

Thermo Scientific Pierce Premium Grade EDC is our highest quality formulation of this popular carbodiimide crosslinker, specially characterized for applications where product integrity and risk minimization are paramount.


  • EDC—carbodiimide crosslinker for conjugation between carboxylates and amines
  • High quality—identity and purity confirmed by several tests
  • Product integrity—enhanced level of testing and characterization compared to standard grade
  • Lot retention—ample supply of past lots retained to ensure future process testing
  • Change management—Change Control Notification (CCN) service
  • Consistent manufacture—batch-specific manufacturing documentation review


Compared to the standard grade product, Premium Grade EDC provides more clearly defined quality and product support by including (a) increased analytical testing and product characterization, (b) greater batch-specific information and quality assurance review, (c) extensive lot sample retention and (d) change control notification. EDC is 1-ethyl-3-[3-dimethylaminopropyl]carbodiimide hydrochloride, a water-soluble carbodiimide crosslinker that activates carboxyl groups for spontaneous reaction with primary amines, enabling peptide immobilization and hapten-carrier protein conjugation. In the presence of N-hydroxysulfosuccinimide (sulfo-NHS), EDC can be used to convert carboxyl groups to amine-reactive sulfo-NHS esters. This is accomplished by mixing the EDC with a carboxyl containing molecule and adding sulfo-NHS.


Specifications of Premium Grade EDC
We manufacture crosslinking reagents to ensure the highest possible overall product integrity, consistency and performance for the intended research applications.
• Visual: White crystalline powder, free of foreign matter
• Identity: IR scan shows only peaks characteristic of EDC
• Water content: Karl Fischer titration, ≤2%
• Trace metals: All metals, including lead, ≤50 ppm
• Elemental analysis: Reported values for C, H, and N


Caution: For Research Use Only. Not for use in diagnostic procedures.

Formula: C₈H₁₇N₃·HCl
Storage Temperature: Freezer
MDL Number: MFCD00012503
CAS Number: 25952-53-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25d8bf2864bcf21ffe6dcca6f22388d1fe427e22&vid=9080dd6b66a1622ef32a2da2138f85e44cc881b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Product Line Pierce™
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Carbodiimide
Spacer Short
Form Solid
Quantity 1 g/25 g/500 g
Type Crosslinker
Solubility Water


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR