Pierce™ HisPur™ Affinity Purification Kits, Thermo Scientific
Supplier: 0000013509
89964 89965 89966 89967 89968 89969 90090 90091 90092
PI89964EA 179.69 USD
PI89964 PI89965 PI89966 PI89967 PI89968 PI89969 PI90090 PI90091 PI90092
Pierce™ HisPur™ Affinity Purification Kits, Thermo Scientific
Protein Purification Protein Purification Kits
Pierce™ HisPur™ Cobalt Purification Kit is designed for the efficient purification of recombinant polyhistidine-tagged proteins from bacterial, mammalian and baculovirus-infected cells.
  • Obtain <10 mg of pure His-tagged protein per mL of resin
  • Resin can be reused several times or discarded after single use
  • Cobalt-chelate coordination core binds fewer host protein contaminants
  • Purify proteins under native or denaturing conditions
His-tagged proteins are purified from total soluble protein extract using a cobalt-charged tetradentate chelator immobilized onto 6% crosslinked agarose. Many immobilized metal affinity chromatography (IMAC) resins contain nickel (Ni2+) for purifying His-tagged proteins. Although Ni2+ chelate resins achieve high protein yields, purity is often sub-optimal, requiring additional clean-up steps. Cobalt achieves both high yield and purity with minimal optimisation. The HisPur™ Cobalt Resin binds fewer non specific proteins, displays less metal leaching and enables less stringent elution conditions compared to Ni2+ resins.
Delivery information: Complete kit contains HisPur™ Cobalt spin columns, pre-formulated Equilibration/Wash and Elution Buffers, collection tubes and tip plugs. Resin and spin columns are also available separately.
Caution: For Research Use Only. Not for use in diagnostic procedures.