Pierce™ Melon™ Gel Purification Buffer, Thermo Scientific
Supplier: 0000013509
89972
PI89972EA 53.98 USD
PI89972
Pierce™ Melon™ Gel Purification Buffer, Thermo Scientific
Buffers Antibody Purification Buffers
Thermo Scientific Melon Gel Purification Buffer, formulation: proprietary buffer dry-blend, sufficient for preparing 200 mL of 100X buffer.
  • Dry powder blend
  • Eliminates need for elution in purification process
  • Time-saving
  • Store between 20˚C and 25˚C
Unlike typical bind-and-elute affinity methods, Melon Gel Purification is based on the binding of non-IgG proteins from a prepared supernatant or ascites sample, thereby allowing the pure IgG to be recovered in the flow-through (non-bound) fraction.
Caution: For Research Use Only. Not for use in diagnostic procedures.