Pierce™ HisPur™ Ni-NTA Spin Purification Kit, Thermo Scientific
Supplier: 0000013509
88227 88228 88229
PI88227PK 531.89 USD
PI88227 PI88228 PI88229
Pierce™ HisPur™ Ni-NTA Spin Purification Kit, Thermo Scientific
Protein Purification Protein Purification Kits
Pierce™ HisPur™ Ni-NTA Spin Purification Kit contains 0.2-ml HisPur™ Ni-NTA spin columns, as well as buffers required to purify polyhistidine-tagged proteins
  • High capacity
  • Compatible
  • Cost-effective
  • Easy to use
  • Flexible
HisPur™ Ni-NTA Resin effectively purifies high levels of overexpressed His-tagged fusion proteins from bacterial lysates, such as those that are attained with Thermo Scientific B-PER Bacterial Protein Extraction Reagents. The resin performs well in batch-binding and spin-column procedures at a variety of scales. Performance equals or exceeds popular Ni-NTA resins from other suppliers. HisPur™ Ni-NTA Resin is a high-quality, stable and resilient affinity support. Tests confirm that no decrease in performance occurs after at least five repeated uses. These data indicated that the resin is highly resistant to structural degradation or nickel ion leaching during normal use.
HisPur™ Ni-NTA resin is a high-capacity, high-performance nickel-IMAC resin for routine affinity purification of His-tagged fusion proteins.
Delivery information: Kit includes HisPur Ni-NTA spin columns, phosphate-buffered saline (10X), imidazole (2 M), collection tubes and column plugs.
Caution: For Research Use Only. Not for use in diagnostic procedures.