Pierce™ HisPur™ Ni-NTA Resin, Thermo Scientific
Supplier: 0000013509
Pierce™ HisPur™ Ni-NTA Resin is a high-capacity, high-performance nickel-IMAC resin for routine affinity purification of His-tagged fusion proteins.
- Binding up to 60 mg of 6xHis-tagged protein per milliliter of resin
- Purify proteins using native or denaturing conditions
- Reuse the same batch of resin at least five times
- Pre-formulated buffers available for kit formats
The specially prepared support consists of beaded agarose derivatized with the nitrilotriacetic acid (NTA) chelation moiety and loaded with divalent nickel ions (Ni²⁺). The immobilized metal affinity chromatography (IMAC) resin provides exceptional binding capacity and performance for recombinant His-tagged protein purification.
Caution: For Research Use Only. Not for use in diagnostic procedures.