Water with 0,1% (v/v) trifluoroacetic acid for LC-MS, Pierce™

Supplier: Thermo Fisher Scientific

T85172-K2
PI85172EA 116.11 USD
PI85172 PI85173
Water with 0,1% (v/v) trifluoroacetic acid for LC-MS, Pierce™
Water with 0,1% (v/v) trifluoroacetic acid

Pierce™ 0.1% Trifluoroacetic Acid (v/v) in Water, LC-MS Grade is a preparation with high purity and low UV-absorptivity that is ideal for HPLC and mass spectrometry applications.


  • Tested more than 20 ways to ensure maximal LC-MS sensitivity
  • Processed, filtered and sealed to extend LC-MS column life
  • Packaged to eliminate variability and reduce handling


0.1% Trifluoroacetic Acid (TFA) in water is specially purified by a proprietary method and tested to ensure lot-to-lot consistency with a low UV absorbance, providing the most sensitive detection across wavelengths and prolonging equipment life. The chromatography- and spectrometry-grade 0.1% TFA in water is 0.2-micron filtered, packaged in solvent-rinsed amber glass bottles and sealed under a nitrogen with PTFE-lined fluorocarbon caps for ultimate protection. For complex separations, the key to success can be to vary selectivity. Varying mobile phase composition on the same column can change selectivity enough to resolve separations that would otherwise overlap. TFA is the most frequently used modifier for separations in reverse-phase HPLC. The usual specified TFA concentration is 0.1%. For reproducible separations from run-to-run or from lab-to-lab, it is essential to make TFA concentrations the same.


Caution: For Research Use Only. Not for use in diagnostic procedures.

Boiling Pt: 71.8 °C (1013 hPa)
Melting Pt: –15 °C
Density: 1.5351 g/cm³ (20 °C)
Storage Temperature: Ambient
ADR: 8,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR