Pierce™ Digestion Indicator for Mass Spectrometry, Thermo Scientific
Supplier: 0000013509
84841
PI84841EA 321.23 USD
PI84841
Pierce™ Digestion Indicator for Mass Spectrometry, Thermo Scientific
Protein Extraction Protein Extraction Reagents
The Digestion Indicator for Mass Spectrometry is a non-mammalian recombinant protein (26kDa) with five signature peptides for use in determining the digestion efficiency and reproducibility across multiple samples.
  • Versatile
  • For use with mass spectrometer
  • Non-mammalian protein source
This product is sufficient for production of five signature peptides upon digestion for mass spectrometry.
Caution: For Research Use Only. Not for use in diagnostic procedures.