Pierce Subcellular Protein Fractionation Kit, Thermo Scientific
Supplier: 0000013509
78840
PI78840EA 736 USD
PI78840
Pierce Subcellular Protein Fractionation Kit, Thermo Scientific
Buffers Extraction Buffers
Thermo Scientific Subcellular Protein Fractionation Kit for Cultured Cells enables segregation and enrichment of proteins from five different cellular compartments.
  • Concise—extract functional cytoplasmic, membrane, nuclear soluble, chromatin-bound and cytoskeletal protein fractions from mammalian cultured cells in less than 3 hours
  • Convenient—perform a simple procedure without using ultracentrifugation over gradients
  • Complete—obtain cytoplasmic, membrane, soluble nuclear, chromatin-bound and cytoskeletal protein fractions from a single kit
  • Cultured Cells—developed and optimized for use with cultured mammalian cells
Fractionation of subcellular proteins enables protein localization assessment and protein enrichment from specific cellular compartments. Thermo Scientific Subcellular Protein Fractionation Kit includes a combination of reagents for stepwise lysis of cells into functional cytoplasmic, membrane, nuclear soluble, chromatin-bound and cytoskeletal protein fractions in less than 3 hours. Extracts from each subcellular compartment generally have less than 15% contamination between fractions, which is sufficient purity for most experiments studying protein localization and redistribution.
Ordering information: Upon receipt store kit at -20 °C, or store individual components as indicated above. Kit is shipped with dry ice.
Caution: For Research Use Only. Not for use in diagnostic procedures.