Perfluoro Acid Anhydrides

Supplier: Thermo Scientific

TS-63164
PI63164EA 393.2 USD
PI63164
Perfluoro Acid Anhydrides
Chromatography Sorbents GC Sorbents

Prepare fluoracyl derivatives for GC/MS and for flame ionization and electron-capture detection techniques with the Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA). These reagents react readily with alcohols, phenols, and amines.


  • Used to prepare fluoracyl derivatives for GC-MS
  • Storage at Room Temperature
  • For use with GC/MS
  • Standard components are Heptafluoro-butanoic acid anhydride
  • Standard Concentration 98 to 100%


Produce stable volatile derivatives for FID and ECD techniques.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR