Pierce™ UltraLink™ Hydrazide Resin Affinity Chromatography Resin, Thermo Scientific
Supplier: 0000013509
53149
PI53149EA 397.54 USD
PI53149
Pierce™ UltraLink™ Hydrazide Resin Affinity Chromatography Resin, Thermo Scientific
Protein Purification Protein Purification Resins
UltraLink™ Hydrazide resin is used to prepare affinity columns with antibodies or other glycolsylated proteins.
  • Able to immobilise 1 to 10 mg of oxidized antibody or other glycoprotein per mL of resin
  • No stabilisation step is required
  • Conjugates only to purified glycoproteins whose sugar groups have been gently oxidised with periodate
  • Immobilise in as few as 30 minutes; achieve at least 90% coupling of most glycoproteins in less than 4 hours (efficiency depends on the amount and type of glycosylation)
Formulation: Hydrazide-activated UltraLink Resin, slurried in water with sodium azide