Toluene-α-sulfonyl fluoride
Supplier: 0000013509
Thermo Scientific PMSF is a protease inhibitor that reacts with serine residues to inhibit trypsin, chymotrypsin, thrombin and papain.
- Targets serine proteases, including trypsin, chymotrypsin, thrombin and papain
- Effective in most protein solutions at 0.1 to 1 mM final concentration
- Because PMSF has limited water solubility, it must be dissolved in a small amount of solvent, such as ethanol, methanol or isopropyl alcohol before addition to a buffer
PMSF is an abbreviation for phenylmethylsulfonyl fluoride, the most common chemical name for this small compound
Properties of PMSF:
Synonyms: alpha-Toluenesulfonyl fluoride; Benzylsulfonyl fluoride; Phenylmethylsulfonyl fluoride
Formula: C6H5CH2SO2F
Molecular weight: 174.19
CAS Number: 329-98-6; EC Number: 206-350-2
R/S Codes: R 25-34; S 26-36/37/39-45
Storage conditions: room temperature in a dry place protected from light
Working solution: dissolve PMSF in isopropanol at 1.74 mg/mL (10 mM); store in aliquots at -20˚C.
Specifications of PMSF (Part No. 36978):
Appearance: white powder
Purity:> 99.0 %
Melting point: 90-95˚C
Humidity: < 0.1 %
In addition to inhibiting serine proteases, PMSF will also inhibit cysteine proteases, like papain (reversible by DTT treatment), and mammalian acetylcholinesterase.
Ordering information: Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25?C.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Formula:
C₇H₇FO₂S |
MDL Number:
MFCD00007424 CAS Number: 329-98-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...