Urea, Pierce™

Supplier: 0000013509

Synonyms: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide

29700
PI29700EA 74.15 USD
PI29700
Urea, Pierce™
Urea

Thermo Scientific Pierce Urea is ≥99% pure, crystalline urea, a protein denaturant with low UV absorptivity. In addition to increasing solubility of hydrophobic molecules, urea is a general protein denaturant, unfolding proteins and altering their three-dimensional structures.


  • Related applications: Protein Labeling & Crosslinking

Formula: NH₂CONH₂
MW: 60.06 g/mol
Boiling Pt: 197 °C (1013 hPa)
Melting Pt: 132.7 °C
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008022
CAS Number: 57-13-6
EINECS: 200-315-5
REACH: 01-2119463277-33
Merck Index: 13,09935

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR