Pierce™ Protein Conjugate, Horseradish Peroxidase, Biotinylated, Thermo Scientific
Supplier: 0000013509
29139
PI29139EA 258.46 USD
PI29139
Pierce™ Protein Conjugate, Horseradish Peroxidase, Biotinylated, Thermo Scientific
Assays Protein Assays
Biotinylated horse radish peroxidase is used as control or signal amplification in IHC to amplify the signal of biotinylated primary antibodies using the avidin-biotin complex staining method.