Pierce PeroXOquant™ Quantitative Peroxide Assay Kits (Aqueous), Thermo Scientific
Supplier: 0000013509
23280
PI23280EA 356.92 USD
PI23280
Pierce PeroXOquant™ Quantitative Peroxide Assay Kits (Aqueous), Thermo Scientific
Assays Protein Assays
Pierce™ Quantitative Peroxide Assay Kit (Aqueous) detects and measures hydrogen peroxide levels in biological samples using an Iron and Xylenol orange* (XO) reagent.
  • Ready to use – removes the variability, difficulty and expense of formulating in-house
  • Ideal for sensitive measurement of peroxide accumulation in non-lipid samples
The Aqueous compatible peroxide assay kit detects peroxides based on oxidation of ferrous to ferric ion in the presence of xylenol orange
Peroxides in the sample oxidize Fe2+ to Fe3+. The Fe3+ ion will form a colored complex with xylenol orange that can be read at 560nm.
*Xylenol orange is (o-cresolsulfonephthalein-3´-3´-bis-[methyliminodiacetic acid sodium salt]).
The kit formulation includes sorbitol, which provides sensitivity enhancement. Most proteins do not interfere with this assay, although some metal chelators may require use of a blank (ie. control) to mitigate these effects.
Delivery information: Aqueous compatible peroxide assay kit includes Reagent A (1 mL) and Reagent B (2x50 mL).