Pierce™ Quantitative Colorimetric Peptide Assay
Supplier: 0000013509
Thermo Scientific™ Pierce™ Quantitative Colorimetric Peptide Assay is an easy-to-use colorimetric microplate assay designed specifically to improve the sensitivity and reproducibility of quantitation of peptide mixtures.
- Sensitive – accurately detect as little as 25 µg/ml of peptide mixture
- Robust – assay performance rigorously tested using peptide digest mixtures
- Robust peptide digest standard – kit includes a validated peptide digest standard for improved reproducibility of quantitation
- Compatible – works with many reagents, including mass spectrometry sample preparation reagents and TMT™-labeled peptides
- Convenient – easy mix-and read-format and stable colorimetric signal that may be read in as little as 15 minutes
The Pierce Quantitative Colorimetric Peptide Assay provides modified BCA reagents for the reduction of Cu+2 to Cu+1 and a proprietary chelator that have been optimized for the quantitation of peptide mixtures. In this reaction, the copper is first reduced by the amide backbone of peptides under alkaline conditions (Biuret reaction). The proprietary chelator then couples with the reduced copper to form a bright red complex with absorbance of 480 nm. The signal produced from this reaction is 3-4 fold more sensitive than the Micro-BCA Protein Assay for peptide analysis.
The kit also contains a high quality peptide digest reference standard for use in generating linear standard curves and to serve as a calibration control. This colorimetric peptide assay requires a small amount of sample (20 µl) and has a working peptide concentration range of 25 to 1000 μg/ml. The increased sensitivity, low sample assay volume, and included reference standard are essential for accurate and robust measurement of peptide digest samples, especially for mass spectrometry applications.
Applications:
Recommended for peptide mixtures
Not recommended for synthetic peptides because the assay is affected by peptide amino acid content
Normalizing sample concentrations for quantitative downstream applications
Normalizing sample loading volumes for LC, MS, and LC/MS applications
Measuring recovery after peptide clean-up procedures
Storage: 2 to 8 °C
Ordering information: Kit contains Colorimettric Assay Reagent A, 50 ml; Colorimetric Assay Reagent B, 2×25 ml; Colorimetric Assay Reagent C, 2 ml; Peptide Digest Assay Standard, 1,5 ml.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...